Anti-EIF3E Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB36766-100EA 930 EUR
ABCAAB36766-100
Anti-EIF3E Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EIF3E (eukaryotic translation initiation factor 3, subunit E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human