Anti-Spi2A Hamster Monoclonal Antibody [clone: MoFo 29.2]

Lieferant: Abcam
ABCAAB36772-50EA 634 EUR
ABCAAB36772-50
Anti-Spi2A Hamster Monoclonal Antibody [clone: MoFo 29.2]
Antikörper
Type: Primary
Antigen: Spi2A
Clonality: monoclonal
Clone: MoFo 29.2
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR