Anti-Vitellogenin Mouse Monoclonal Antibody [clone: ND-1E8]

Lieferant: Abcam
ABCAAB36795-50EA 690 EUR
ABCAAB36795-50
Anti-Vitellogenin Mouse Monoclonal Antibody [clone: ND-1E8]
Antikörper
Type: Primary
Antigen: Vitellogenin
Clonality: monoclonal
Clone: ND-1E8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR