Anti-Vitellogenin Mouse Monoclonal Antibody [clone: JE-10D4]

Lieferant: Abcam
ABCAAB36804-50EA 672 EUR
ABCAAB36804-50
Anti-Vitellogenin Mouse Monoclonal Antibody [clone: JE-10D4]
Antikörper
Type: Primary
Antigen: Vitellogenin
Clonality: monoclonal
Clone: JE-10D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR