Anti-ATM Mouse monoclonal antibody unconjugated [clone: 10H11.E12]

Lieferant: Abcam
ABCAAB36810-100EA 719 EUR
ABCAAB36810-100
Anti-ATM Mouse monoclonal antibody unconjugated [clone: 10H11.E12]
Antikörper
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: monoclonal
Clone: 10H11.E12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR