Anti-TP53BP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB36823-100EA 719 EUR
ABCAAB36823-100
Anti-TP53BP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TP53BP1 (tumor protein p53 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR