Anti-CCR3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB36827-100EA 688 EUR
ABCAAB36827-100
Anti-CCR3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CCR3 (chemokine (C-C motif) receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR