Anti-CARD10 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB36839-100EA 878 EUR
ABCAAB36839-100
Anti-CARD10 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CARD10 (caspase recruitment domain family, member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR