Anti-Vitellogenin Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB36877-100EA 882 EUR
ABCAAB36877-100
Anti-Vitellogenin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Vitellogenin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen