Anti-C1QTNF5 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB36893-100EA 856 EUR
ABCAAB36893-100
Anti-C1QTNF5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C1QTNF5 (C1q and tumor necrosis factor related protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat