Anti-NTRK1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB36961-100EA 672 EUR
ABCAAB36961-100
Anti-NTRK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NTRK1 (neurotrophic tyrosine kinase, receptor, type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR