Anti-TRAF3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB36988-100EA 682 EUR
ABCAAB36988-100
Anti-TRAF3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRAF3 (TNF receptor-associated factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR