Anti-MAPK1/3 Mouse Monoclonal Antibody [clone: 9B3]

Lieferant: Abcam
ABCAAB36991-100EA 697 EUR
ABCAAB36991-100
Anti-MAPK1/3 Mouse Monoclonal Antibody [clone: 9B3]
Antikörper
Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: monoclonal
Clone: 9B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR