Anti-SMARCA1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37003-100EA 713 EUR
ABCAAB37003-100
Anti-SMARCA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMARCA1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR