Anti-TLR1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37068-100EA 688 EUR
ABCAAB37068-100
Anti-TLR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TLR1 (toll-like receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR