Anti-BCAP31 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB37120-100EA 697 EUR
ABCAAB37120-100
Anti-BCAP31 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BCAP31 (B-cell receptor-associated protein 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR