Anti-Cytochrome P450 1A1 + 1A2 Mouse Monoclonal Antibody [clone: C10-7]

Lieferant: Abcam
ABCAAB37130-100EA 690 EUR
ABCAAB37130-100
Anti-Cytochrome P450 1A1 + 1A2 Mouse Monoclonal Antibody [clone: C10-7]
Antikörper
Type: Primary
Antigen: Cytochrome P450 1A1 + 1A2
Clonality: monoclonal
Clone: C10-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR