Anti-Cytochrome P450 1A1 + 1A2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37131-100EA 690 EUR
ABCAAB37131-100
Anti-Cytochrome P450 1A1 + 1A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Cytochrome P450 1A1 + 1A2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR