Anti-DDX3X Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB37160-100EA 895 EUR
ABCAAB37160-100
Anti-DDX3X Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DDX3X (DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat