Anti-KDM1A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37165-100EA 906 EUR
ABCAAB37165-100
Anti-KDM1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KDM1A (lysine (K)-specific demethylase 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR