Anti-GAPDH Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB37168-100EA 947 EUR
ABCAAB37168-100
Anti-GAPDH Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen