Anti-TNFSF15 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37169-100EA 666 EUR
ABCAAB37169-100
Anti-TNFSF15 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNFSF15 (tumor necrosis factor (ligand) superfamily, member 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR