Anti-CLU Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37172-100EA 666 EUR
ABCAAB37172-100
Anti-CLU Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLU (clusterin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR