Anti-SIGN Related 1 Rat Monoclonal Antibody [clone: ER-TR9]

Lieferant: Abcam
ABCAAB37220-100EA 703 EUR
ABCAAB37220-100
Anti-SIGN Related 1 Rat Monoclonal Antibody [clone: ER-TR9]
Antikörper
Type: Primary
Antigen: SIGN Related 1
Clonality: monoclonal
Clone: ER-TR9
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR