Anti-CHIPS Mouse Monoclonal Antibody [clone: JNC1]

Lieferant: Abcam
ABCAAB37241-50EA 634 EUR
ABCAAB37241-50
Anti-CHIPS Mouse Monoclonal Antibody [clone: JNC1]
Antikörper
Type: Primary
Antigen: CHIPS
Clonality: monoclonal
Clone: JNC1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR