Anti-CTSK Mouse Monoclonal Antibody [clone: 3F9]

Lieferant: Abcam
ABCAAB37259-50EA 703 EUR
ABCAAB37259-50
Anti-CTSK Mouse Monoclonal Antibody [clone: 3F9]
Antikörper
Type: Primary
Antigen: CTSK (cathepsin K)
Clonality: monoclonal
Clone: 3F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR