Anti-Cholecystokinin 8 Mouse Monoclonal Antibody [clone: HYB 345-02]
Lieferant: Abcam
ABCAAB37274-100EA 1010 EUR
ABCAAB37274-100
Anti-Cholecystokinin 8 Mouse Monoclonal Antibody [clone: HYB 345-02]
Antikörper
Type: Primary
Antigen: Cholecystokinin 8
Clonality: monoclonal
Clone: HYB 345-02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat