Anti-PPY Mouse Monoclonal Antibody [clone: HYB 347-06]

Lieferant: Abcam
ABCAAB37285-100EA 666 EUR
ABCAAB37285-100
Anti-PPY Mouse Monoclonal Antibody [clone: HYB 347-06]
Antikörper
Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: monoclonal
Clone: HYB 347-06
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR