Anti-HAGH Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB37432-50EA 792 EUR
ABCAAB37432-50
Anti-HAGH Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HAGH (hydroxyacylglutathione hydrolase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR