Anti-LSM1 Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB37463-50EA 911 EUR
ABCAAB37463-50
Anti-LSM1 Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LSM1 (LSM1, U6 small nuclear RNA associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR