Anti-SNAPIN Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB37496-50EA 758 EUR
ABCAAB37496-50
Anti-SNAPIN Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNAPIN (SNAP-associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR