Anti-U2AF2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37530-200EA 719 EUR
ABCAAB37530-200
Anti-U2AF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: U2AF2 (U2 small nuclear RNA auxiliary factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR