Anti-CRAT Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB37588-100EA 722 EUR
ABCAAB37588-100
Anti-CRAT Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CRAT (carnitine O-acetyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR