Anti-Staphylococcus aureus Mouse Monoclonal Antibody [clone: 704]

Lieferant: Abcam
ABCAAB37644-100EA 728 EUR
ABCAAB37644-100
Anti-Staphylococcus aureus Mouse Monoclonal Antibody [clone: 704]
Antikörper
Type: Primary
Antigen: Staphylococcus aureus
Clonality: monoclonal
Clone: 704
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR