Anti-S100A12 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37657-200EA 688 EUR
ABCAAB37657-200
Anti-S100A12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S100A12 (S100 calcium binding protein A12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR