Anti-Bordetella pertussis toxin Mouse monoclonal antibody unconjugated [clone: 56.2]

Lieferant: Abcam
ABCAAB37686-100EA 666 EUR
ABCAAB37686-100
Anti-Bordetella pertussis toxin Mouse monoclonal antibody unconjugated [clone: 56.2]
Antikörper
Type: Primary
Antigen: Bordetella pertussis toxin
Clonality: monoclonal
Clone: 56.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR