Anti-SGTA Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB37706-50EA 763 EUR
ABCAAB37706-50
Anti-SGTA Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SGTA (small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR