Anti-ADAMTS12 Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB37760-50EA 750 EUR
ABCAAB37760-50
Anti-ADAMTS12 Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAMTS12 (ADAM metallopeptidase with thrombospondin type 1 motif, 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR