Anti-C1QBP Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB37851-50EA 672 EUR
ABCAAB37851-50
Anti-C1QBP Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C1QBP (complement component 1, q subcomponent binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR