Anti-ADI1 Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB37877-50EA 750 EUR
ABCAAB37877-50
Anti-ADI1 Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADI1 (acireductone dioxygenase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR