Anti-NDRG1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB37897-100EA 902 EUR
ABCAAB37897-100
Anti-NDRG1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NDRG1 (N-myc downstream regulated 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR