Anti-C12orf5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB37910-100EA 713 EUR
ABCAAB37910-100
Anti-C12orf5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C12orf5 (chromosome 12 open reading frame 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR