Anti-ARPC3 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB37916-100EA 634 EUR
ABCAAB37916-100
Anti-ARPC3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARPC3 (actin related protein 2/3 complex, subunit 3, 21kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR