Anti-UBE2K Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB37917-100EA 697 EUR
ABCAAB37917-100
Anti-UBE2K Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBE2K (ubiquitin-conjugating enzyme E2K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR