Anti-eEF1A1+eEF1A2+eEF1AL3 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB37969-100EA 929 EUR
ABCAAB37969-100
Anti-eEF1A1+eEF1A2+eEF1AL3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: eEF1A1+eEF1A2+eEF1AL3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat