Anti-VDAC2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB37985-100EA 713 EUR
ABCAAB37985-100
Anti-VDAC2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VDAC2 (voltage-dependent anion channel 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR