Anti-RBBP4 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB38135-100EA 892 EUR
ABCAAB38135-100
Anti-RBBP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RBBP4 (retinoblastoma binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human