Anti-TFRC Mouse Monoclonal Antibody [clone: 13E4]

Lieferant: Abcam
ABCAAB38171-500EA 688 EUR
ABCAAB38171-500
Anti-TFRC Mouse Monoclonal Antibody [clone: 13E4]
Antikörper
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: monoclonal
Clone: 130000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR