Anti-Adenylate cyclase 1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38331-50EA 672 EUR
ABCAAB38331-50
Anti-Adenylate cyclase 1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Adenylate cyclase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR