Anti-DNA polymerase delta p50 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38338-100EA 891 EUR
ABCAAB38338-100
Anti-DNA polymerase delta p50 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNA polymerase delta p50
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR